accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 872041-85-5

IUPAC Name: (5-chloropyridin-3-yl)boronic acid
Synonyms: 5-chloropyridin-3-ylboronic acid
Molecular Weight (g/mol): 157.36
Molecular Formula: C5H5BClNO2
InChi Key: NJXYBTMCTZAUEE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CN=C1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results