accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BONONCH3

CAS RN 872054-54-1

IUPAC Name: 2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane
Synonyms: 2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane
Molecular Weight (g/mol): 282.15
Molecular Formula: C16H19BN2O2
InChi Key: AVMWJACSYAQNDG-UHFFFAOYSA-N
SMILES: CC1=CN=C(C=C1)B1OCCN(CCO1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results