CAS RN 87226-41-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-(-)-Etodolac, TRC
CAS: 87226-41-3 Molecular Formula: C17 H21 N O3 Molecular Weight (g/mol): 287.354 Synonym: (1R)-1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic Acid,(-)-(R)-Etodolac,(-)-Etodolac,(-)-Etodolic Acid,R-Etodolac,RAK 593,SDX 101 IUPAC Name: 2-[(1R)-1,8-diethyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl]acetic acid SMILES: CCc1cccc2c3CCO[C@](CC)(CC(=O)O)c3[nH]c12