accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNNH2O

CAS RN 873-50-7

IUPAC Name: 3-methyl-1H-pyrazole-1-carboxamide
Synonyms: 1-carbamoyl-3-methylpyrazole
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: PNZZQMXKKNDHNS-UHFFFAOYSA-N
SMILES: CC1=NN(C=C1)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results