accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 873305-35-2

OHNONN

CAS RN 873305-35-2

IUPAC Name: N-[(oxolan-2-yl)methyl]-5,6-diphenylfuro[2,3-d]pyrimidin-4-amine
Synonyms: N-(oxolan-2-ylmethyl)-5,6-diphenylfuro[2,3-d]pyrimidin-4-amine
Molecular Weight (g/mol): 371.44
Molecular Formula: C23H21N3O2
InChi Key: XNFHHOXCDUAYSR-UHFFFAOYNA-N
SMILES: C(NC1=C2C(OC(=C2C2=CC=CC=C2)C2=CC=CC=C2)=NC=N1)C1CCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results