accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNHNCl

CAS RN 87394-55-6

IUPAC Name: 1-(3-chloropyridin-2-yl)piperazine
Synonyms: 1-(3-chloropyridin-2-yl)piperazine
Molecular Weight (g/mol): 197.67
Molecular Formula: C9H12ClN3
InChi Key: HLCPXCHNWZGOMT-UHFFFAOYSA-N
SMILES: ClC1=C(N=CC=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results