CAS RN 874-14-6
IUPAC Name:
1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
1,3-dimethyluracil N,N'-dimethyluracil N,n'-dimethyluracil
Molecular Weight (g/mol):
140.14
Molecular Formula:
C6H8N2O2
InChi Key:
JSDBKAHWADVXFU-UHFFFAOYSA-N
SMILES:
CN1C=CC(=O)N(C)C1=O
1
–
1
of
1
results
1
1,3-Dimethyluracil, 99%, Thermo Scientific Chemicals
CAS: 874-14-6 Molecular Formula: C6H8N2O2 Molecular Weight (g/mol): 140.14 MDL Number: MFCD00038065 InChI Key: JSDBKAHWADVXFU-UHFFFAOYSA-N Synonym: 1,3-dimethyluracil,n1,n3-dimethyluracil,n,n'-dimethyluracil,1,3-dimethylpyrimidine-2,4 1h,3h-dione,2,4 1h,3h-pyrimidinedione, 1,3-dimethyl,2,4-dihydroxy-1,3-dimethylpyrimidine,uracil, 1,3-dimethyl,1,3-dimethyl-2,4 1h,3h-pyrimidinedione,1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione,1,3-dimethyl-1h-pyrimidine-2,4-dione PubChem CID: 70122 ChEBI: CHEBI:74763 IUPAC Name: 1,3-dimethylpyrimidine-2,4-dione SMILES: CN1C=CC(=O)N(C)C1=O