accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 874-18-0

NH2ClBrBr

CAS RN 874-18-0

IUPAC Name: 2,4-dibromo-6-chloroaniline
Synonyms: 2,4-dibromo-6-chloroaniline
Molecular Weight (g/mol): 285.36
Molecular Formula: C6H4Br2ClN
InChi Key: MFROLNRQDCAZCD-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results