accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOIOH3COOCH3OOO

CAS RN 87413-09-0

IUPAC Name: 1,1-bis(acetyloxy)-3-oxo-3H-1λ⁵,2-benziodaoxol-1-yl acetate
Synonyms: dess-martin periodinane
Molecular Weight (g/mol): 424.14
Molecular Formula: C13H13IO8
InChi Key: NKLCNNUWBJBICK-UHFFFAOYSA-N
SMILES: CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)