accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BONCH3H3CHOOH

CAS RN 874219-16-6

IUPAC Name: [2-(dimethylcarbamoyl)phenyl]boronic acid
Synonyms: 2-(dimethylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 193.01
Molecular Formula: C9H12BNO3
InChi Key: NZIOVLXULSCCSG-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results