accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 874288-05-8

BOHNFHOOH

CAS RN 874288-05-8

IUPAC Name: {4-[(3-fluorophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 4-[(3-fluorophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 259.04
Molecular Formula: C13H11BFNO3
InChi Key: CVYDMRNYGKMSDI-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)NC1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results