accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 874473-14-0

IUPAC Name: 3-methyloxetan-3-amine
Synonyms: 3-methyloxetan-3-amine
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: NQVWMPOQWBDSAI-UHFFFAOYSA-N
SMILES: CC1(N)COC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results