accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 875-51-4

IUPAC Name: 4-bromo-2-nitroaniline
Synonyms: benzenamine, 4-bromo-2-nitro-
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: ZCWBZRBJSPWUPG-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)