accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87583-89-9

H3CNHHNCH3

CAS RN 87583-89-9

IUPAC Name: N1,N2-dimethylcyclohexane-1,2-diamine
Synonyms: N1,N2-dimethylcyclohexane-1,2-diamine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: JRHPOFJADXHYBR-UHFFFAOYNA-N
SMILES: CNC1CCCCC1NC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results