accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 876-08-4

ClClO

CAS RN 876-08-4

IUPAC Name: 4-(chloromethyl)benzoyl chloride
Synonyms: 4-(chloromethyl)benzoyl chloride
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: RCOVTJVRTZGSBP-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(C=C1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)