accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87600-98-4

NNClOOHCl

CAS RN 87600-98-4

IUPAC Name: 4,6-dichloropyrimidine-5-carboxylic acid
Synonyms: 4,6-dichloropyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 192.98
Molecular Formula: C5H2Cl2N2O2
InChi Key: KGBLYFGHJNGQBK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)N=CN=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results