accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNBrCH3

CAS RN 876728-35-7

IUPAC Name: 6-bromo-1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline
Synonyms: 6-bromo-1,4-dimethyl-2,3-dihydroquinoxaline
Molecular Weight (g/mol): 241.13
Molecular Formula: C10H13BrN2
InChi Key: BVWZHBZUOICVIU-UHFFFAOYSA-N
SMILES: CN1CCN(C)C2=C1C=CC(Br)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results