accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 876728-41-5

IUPAC Name: [5-(furan-2-yl)-1-methyl-1H-pyrazol-3-yl]methanol
Synonyms: [5-(furan-2-yl)-1-methylpyrazol-3-yl]methanol
Molecular Weight (g/mol): 178.19
Molecular Formula: C9H10N2O2
InChi Key: XKIFRVDOWHFUHG-UHFFFAOYSA-N
SMILES: CN1N=C(CO)C=C1C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results