accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87674-68-8

IUPAC Name: 2-chloro-N-(2,4-dimethylthiophen-3-yl)-N-(1-methoxypropan-2-yl)acetamide
Synonyms: dimethenamid
Molecular Weight (g/mol): 275.79
Molecular Formula: C12H18ClNO2S
InChi Key: JLYFCTQDENRSOL-UHFFFAOYNA-N
SMILES: COCC(C)N(C(=O)CCl)C1=C(C)SC=C1C
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results