accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONOO

CAS RN 87687-03-4

IUPAC Name: 7-(pentyloxy)-3H-phenoxazin-3-one
Synonyms: 7-(pentyloxy)phenoxazin-3-one
Molecular Weight (g/mol): 283.33
Molecular Formula: C17H17NO3
InChi Key: ZPSOKQFFOYYPKC-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C2N=C3C=CC(=O)C=C3OC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results