CAS RN 877-24-7
Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical™
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 MDL Number: 13070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.22 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, 99+%
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.22 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, 99%
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, 99.99%, acidimetric standard
CAS: 877-24-7 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
pH Buffer Pouches, Oakton™
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, 0.05N Standardized Solution
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, for HPLC
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.22 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M IUPAC Name: potassium 2-carboxybenzoate SMILES: [K+].OC(=O)C1=CC=CC=C1C([O-])=O
Potassium Phthalate Monobasic, ≥99.5% (T), For HPLC, Solstice
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Standard Buffer Phosphate, pH 4.002, NIST, Chem-Lab
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium Biphthalate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
- 1
- 2