CAS RN 87777-46-6
IUPAC Name:
4-methylhept-1-yn-3-ol
Synonyms:
4-methylhept-1-yn-3-ol
Molecular Weight (g/mol):
126.2
Molecular Formula:
C8H14O
InChi Key:
WONUPNLSAKXVSN-UHFFFAOYNA-N
SMILES:
CCCC(C)C(O)C#C
1
–
1
of
1
results
1
4-Methyl-1-heptyn-3-ol, 97%, Thermo Scientific™
CAS: 87777-46-6 Molecular Formula: C8H14O Molecular Weight (g/mol): 126.199 MDL Number: MFCD00041639 InChI Key: WONUPNLSAKXVSN-UHFFFAOYSA-N Synonym: 4-methyl-1-heptyn-3-ol,4-methyl-1-heptyne-3-ol,4-methyl-hept-1-yn-3-ol,1-heptyn-3-ol,4-methyl PubChem CID: 20442077 IUPAC Name: 4-methylhept-1-yn-3-ol SMILES: CCCC(C)C(C#C)O