accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FFFO

CAS RN 878001-20-8

IUPAC Name: 2-methyl-3-(trifluoromethyl)benzaldehyde
Synonyms: 2-methyl-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: CUQHTBQRXZCFTC-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results