accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFNOOOOH

CAS RN 878572-17-9

IUPAC Name: 4-fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
Synonyms: 4-fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 253.11
Molecular Formula: C8H3F4NO4
InChi Key: XCZRYPKVIJEHSU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=C(F)C(=C1)[N+]([O-])=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results