accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 879636-95-0

CH3SNOH3COOH

CAS RN 879636-95-0

IUPAC Name: 2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: 2-(3-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 249.28
Molecular Formula: C12H11NO3S
InChi Key: WFBWIROXKGGXAW-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1=NC(C)=C(S1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results