accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2CNO

CAS RN 88-12-0

IUPAC Name: 1-ethenylpyrrolidin-2-one
Synonyms: N-vinylpyrrolidone polyvinylpolypyrrolidone PVP
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N
SMILES: C=CN1CCCC1=O
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)