accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88-16-4

FFFCl

CAS RN 88-16-4

IUPAC Name: 1-chloro-2-(trifluoromethyl)benzene
Synonyms: chloro(trifluoromethyl)benzene
Molecular Weight (g/mol): 180.55
Molecular Formula: C7H4ClF3
InChi Key: DGRVQOKCSKDWIH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results