accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88-19-7

CH3SH2NOO

CAS RN 88-19-7

IUPAC Name: 2-methylbenzene-1-sulfonamide
Synonyms: O-methylbenzenesulfonamide
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: YCMLQMDWSXFTIF-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1S(N)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)