accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2ClSOHOO

CAS RN 88-51-7

IUPAC Name: 2-amino-4-chloro-5-methylbenzene-1-sulfonic acid
Synonyms: 2-amino-4-chloro-5-methylbenzenesulfonic acid
Molecular Weight (g/mol): 221.66
Molecular Formula: C7H8ClNO3S
InChi Key: VRLPHBSFRWMMPW-UHFFFAOYSA-N
SMILES: CC1=CC(=C(N)C=C1Cl)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)