accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88-58-4

CH3H3CCH3OHHOCH3H3CCH3

CAS RN 88-58-4

IUPAC Name: 2,5-di-tert-butylbenzene-1,4-diol
Synonyms: di-T-butylhydroquinone
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: JZODKRWQWUWGCD-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)