accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88-69-7

IUPAC Name: 2-(propan-2-yl)phenol
Synonyms: 2-isopropylphenol prodox 131
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: CRBJBYGJVIBWIY-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)