accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOOHO

CAS RN 88-97-1

IUPAC Name: 2-carbamoylbenzoic acid
Synonyms: phthalamic acid phthalamide acid phthalic monoamide phthalamidic acid
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: CYMRPDYINXWJFU-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results