accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHO

CAS RN 88-99-3

IUPAC Name: benzene-1,2-dicarboxylic acid
Synonyms: phthalic acid sunftal 20 o-phthalic acid kyselina ftalova alizarinic acid naphthalinate alizarinate ortho-phthalic acid acide phtalique O-phthalic acid
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: XNGIFLGASWRNHJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (6)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (1)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (2)