accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNOO

CAS RN 881-03-8

IUPAC Name: 2-methyl-1-nitronaphthalene
Synonyms: naphthalene, 2-methyl-1-nitro-
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: IZNWACYOILBFEG-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results