accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 881-86-7

CH3OONOOCH3

CAS RN 881-86-7

IUPAC Name: 2,5-dimethyl pyridine-2,5-dicarboxylate
Synonyms: 2,5-dimethyl pyridine-2,5-dicarboxylate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: TUGSJNQAIMFEDY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N=C1)C(=O)OC
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)