accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88105-17-3

CH3OOSCl

CAS RN 88105-17-3

IUPAC Name: methyl 3-chlorothiophene-2-carboxylate
Synonyms: methyl 3-chlorothiophene-2-carboxylate
Molecular Weight (g/mol): 176.61
Molecular Formula: C6H5ClO2S
InChi Key: ZWKYYONTMGVPSP-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C=CS1
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)