accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 881189-65-7

IUPAC Name: 3-(4-chloro-3-fluorophenyl)propanoic acid
Synonyms: 3-(4-chloro-3-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: GDXMVKNNJGWZFN-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(F)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results