accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88189-03-1

FFFSOOOFFFSOOOFFFSOOOBi

CAS RN 88189-03-1

IUPAC Name: bismuth(3+) tritrifluoromethanesulfonate
Synonyms: bismuth(3+) tritriflate
Molecular Weight (g/mol): 656.17
Molecular Formula: C3BiF9O9S3
InChi Key: NYENCOMLZDQKNH-UHFFFAOYSA-K
SMILES: [Bi+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results