accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88192-19-2

IUPAC Name: 3-azidopropan-1-amine
Synonyms: 3-azidopropan-1-amine
Molecular Weight (g/mol): 100.13
Molecular Formula: C3H8N4
InChi Key: OYBOVXXFJYJYPC-UHFFFAOYSA-N
SMILES: NCCCN=[N+]=[N-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results