accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONSNF

CAS RN 881970-80-5

IUPAC Name: 3-{2-[(4-fluorophenyl)methyl]imidazo[2,1-b][1,3]thiazol-6-yl}-2H-chromen-2-one
Synonyms: 3-{2-[(4-fluorophenyl)methyl]imidazo[2,1-b][1,3]thiazol-6-yl}chromen-2-one
Molecular Weight (g/mol): 376.41
Molecular Formula: C21H13FN2O2S
InChi Key: IWFKQTWYILKFGE-UHFFFAOYSA-N
SMILES: FC1=CC=C(CC2=CN3C=C(N=C3S2)C2=CC3=CC=CC=C3OC2=O)C=C1
Product Line 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results