accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 882678-96-8

IUPAC Name: 2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 233.12
Molecular Formula: C13H20BNO2
InChi Key: JMKMGPGFYMANCA-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1N)B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results