CAS RN 883-93-2
IUPAC Name:
2-phenyl-1,3-benzothiazole
Synonyms:
2-phenylbenzthiazole
Molecular Weight (g/mol):
211.28
Molecular Formula:
C13H9NS
InChi Key:
XBHOUXSGHYZCNH-UHFFFAOYSA-N
SMILES:
S1C2=CC=CC=C2N=C1C1=CC=CC=C1
1
–
2
of
2
results
1
2-Phenylbenzothiazole, TRC
CAS: 883-93-2 Molecular Formula: C13H9NS Molecular Weight (g/mol): 211.28 Synonym: 2-Phenylbenzothiazole,Benzothiazole, 2-phenyl-,2-Phenylbenzothiazole,2-Phenyl-1,3-benzothiazole,2-Phenylbenzo[d]thiazole,NSC 1854,NSC 2034 IUPAC Name: 2-phenyl-1,3-benzothiazole SMILES: c1ccc(cc1)c2nc3ccccc3s2