accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 883-99-8

H3COOHO

CAS RN 883-99-8

IUPAC Name: methyl 3-hydroxynaphthalene-2-carboxylate
Synonyms: methyl 3-hydroxynaphthalene-2-carboxylate
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: YVVBECLPRBAATK-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C=C2C=CC=CC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)