accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHO

CAS RN 884-33-3

IUPAC Name: 2-benzylbutanedioic acid
Synonyms: benzylsuccinic acid β-carboxybenzenebutanoic acid
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: GTOFKXZQQDSVFH-UHFFFAOYNA-N
SMILES: OC(=O)CC(CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)