accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOBrOH3C

CAS RN 88404-25-5

IUPAC Name: 4-(bromomethyl)-6,7-dimethoxy-2H-chromen-2-one
Synonyms: 4-(bromomethyl)-6,7-dimethoxychromen-2-one
Molecular Weight (g/mol): 299.12
Molecular Formula: C12H11BrO4
InChi Key: JGODLBJJCNQFII-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(CBr)=CC(=O)OC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results