accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88416-50-6

IUPAC Name: disodium tetrahydrate dihydrogen [dichloro(hydrogen phosphonato)methyl]phosphonate
Synonyms: disodium tetrahydrate bis(clondronate(2-))
Molecular Weight (g/mol): 603.78
Molecular Formula: C2H12Cl4Na2O16P4
InChi Key: MSZARZDVBJBTIF-UHFFFAOYSA-J
SMILES: O.O.O.O.[Na+].[Na+].OP([O-])(=O)C(Cl)(Cl)P(O)([O-])=O.OP([O-])(=O)C(Cl)(Cl)P(O)([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results