CAS RN 884507-45-3
CAS RN 884507-45-3
IUPAC Name:
1-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidine
Synonyms:
1-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidine
Molecular Weight (g/mol):
287.21
Molecular Formula:
C17H26BNO2
InChi Key:
AKQLICZDOCQKRA-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC(CN2CCCC2)=CC=C1
1
–
1
of
1
results
1
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine, 97%, Thermo Scientific™
CAS: 884507-45-3 Molecular Formula: C17H26BNO2 Molecular Weight (g/mol): 287.21 MDL Number: MFCD08741425 InChI Key: AKQLICZDOCQKRA-UHFFFAOYSA-N Synonym: 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl pyrrolidine,1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrolidine,amtb769,3-pyrrolidinomethyl phenylboronic acid, pinacol ester,3-pyrrolidine methyl phenylboronic acid pinacol ester,pyrrolidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl PubChem CID: 18525872 SMILES: CC1(C)OB(OC1(C)C)C1=CC(CN2CCCC2)=CC=C1