accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNOH

CAS RN 884507-50-0

IUPAC Name: {4-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl}methanol
Synonyms: {4-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl}methanol
Molecular Weight (g/mol): 234.34
Molecular Formula: C14H22N2O
InChi Key: YQUWAYPNHCWYNU-UHFFFAOYSA-N
SMILES: CN1CCCN(CC2=CC=C(CO)C=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results