CAS RN 88462-46-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(8-Carboxyoctyl)triphenylphosphonium Bromide, TRC
CAS: 88462-46-8 Molecular Formula: C27H32BrO2P Molecular Weight (g/mol): 499.42 Synonym: 9-(Bromotriphenylphosphoranyl)nonanoic Acid,(8-Carboxyoctyl)triphenyl-phosphonium Bromide IUPAC Name: 8-carboxyoctyl(triphenyl)phosphanium;bromide SMILES: OC(=O)CCCCCCCCP(Br)(c1ccccc1)(c2ccccc2)c3ccccc3