accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88497-27-2

IUPAC Name: 6-bromopyridazin-3-amine
Synonyms: 6-bromopyridazin-3-amine
Molecular Weight (g/mol): 174
Molecular Formula: C4H4BrN3
InChi Key: FXTDHDQFLZNYKW-UHFFFAOYSA-N
SMILES: NC1=NN=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results